Assay Details
| Gene Symbol: Setd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040398.2 | NP_001035488.2 | 8 - 9 | 3229 | 76 | |
| GenBank mRNA | BC038367.1 | - | 2 - 3 | 307 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Setd1b
Genomic Map