Assay Details
| Gene Symbol: Atl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286647.1 | NP_001273576.1 | 10 - 11 | 1055 | 71 | |
| NM_019717.3 | NP_062691.3 | 11 - 12 | 1300 | 71 | ||
| NM_178050.4 | NP_835151.2 | 10 - 11 | 1055 | 71 | ||
| GenBank mRNA | AK032050.1 | - | 2 - 3 | 1651 | 71 | |
| AK038697.1 | - | 10 - 11 | 983 | 71 | ||
| AK135988.1 | - | 10 - 11 | 946 | 71 | ||
| BC006934.1 | - | 3 - 4 | 183 | 71 | ||
| BC051095.1 | - | 2 - 3 | 124 | 71 | ||
| BC055466.1 | - | 8 - 9 | 658 | 71 | ||
| BC060501.1 | - | 11 - 12 | 1205 | 5362692 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atl2
Genomic Map