Assay Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081006.1 | NP_001074475.1 | 12 - 13 | 2561 | 70 | |
| NM_001177630.2 | NP_001171101.2 | 11 - 12 | 2659 | 70 | ||
| NM_001177631.1 | NP_001171102.1 | 10 - 11 | 2554 | 70 | ||
| NM_029895.4 | NP_084171.2 | 11 - 12 | 2767 | 70 | ||
| NM_178059.5 | NP_835160.2 | 11 - 12 | 2767 | 70 | ||
| GenBank mRNA | AB093289.2 | - | 10 - 11 | 2344 | 70 | |
| AB125594.2 | - | 11 - 12 | 2767 | 70 | ||
| AB125595.2 | - | 11 - 12 | 2767 | 70 | ||
| AK054453.1 | - | 6 - 7 | 1710 | 70 | ||
| AK086078.1 | - | 6 - 7 | 1712 | 70 | ||
| AK147510.1 | - | 10 - 11 | 2461 | 70 | ||
| BC158051.1 | - | 11 - 12 | 2381 | 9007405 | 70 | |
| BC172143.1 | - | 10 - 11 | 2274 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Etl4
Genomic Map