Assay Details
| Gene Symbol: Cox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178379.3 | NP_848466.1 | 5 - 6 | 785 | 70 | |
| GenBank mRNA | AK149999.1 | - | 5 - 6 | 775 | 70 | |
| AK154668.1 | - | 5 - 6 | 784 | 70 | ||
| AK171661.1 | - | 5 - 6 | 775 | 70 | ||
| AK178210.1 | - | 2 - 3 | 107 | 70 | ||
| AK186700.1 | - | 2 - 3 | 107 | 70 | ||
| BC018301.1 | - | 5 - 6 | 736 | 70 | ||
| BC038046.1 | - | 5 - 6 | 738 | 4206826 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cox10
Genomic Map