Assay Details
| Gene Symbol: Myo1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178440.4 | NP_848534.2 | 18 - 19 | 2652 | 62 | |
| GenBank mRNA | AK030825.1 | - | 18 - 19 | 2653 | 62 | |
| AK036461.1 | - | 18 - 19 | 2637 | 62 | ||
| AK042033.1 | - | 18 - 19 | 2605 | 62 | ||
| AK088011.1 | - | 18 - 19 | 2653 | 62 | ||
| AK152797.1 | - | 18 - 19 | 2646 | 62 | ||
| AK159700.1 | - | 18 - 19 | 2628 | 62 | ||
| AK171284.1 | - | 18 - 19 | 2646 | 62 | ||
| AK171505.1 | - | 18 - 19 | 2626 | 62 | ||
| BC028661.1 | - | 6 - 7 | 940 | 62 | ||
| BC113755.1 | - | 18 - 19 | 2596 | 40061202 | 62 | |
| BC113954.1 | - | 18 - 19 | 2596 | 40061200 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myo1g
Genomic Map