Assay Details
| Gene Symbol: Mfsd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178620.3 | NP_848735.1 | 2 - 3 | 215 | 91 | |
| GenBank mRNA | AF015191.1 | - | 2 - 3 | 181 | 91 | |
| AK031384.1 | - | 2 - 3 | 410 | 91 | ||
| AK032043.1 | - | 2 - 3 | 411 | 91 | ||
| AK080006.1 | - | 2 - 3 | 937 | 91 | ||
| BC016209.1 | - | 2 - 3 | 199 | 91 | ||
| BC079583.1 | - | 2 - 3 | 239 | 91 | ||
| BC088988.1 | - | 2 - 3 | 215 | 6418757 | 91 | |
| BC100547.1 | - | 3 - 4 | 372 | 91 | ||
| BC137970.1 | - | 2 - 3 | 202 | 8860989 | 91 | |
| BC137971.1 | - | 2 - 3 | 203 | 8860990 | 91 | |
| BC145068.1 | - | 2 - 3 | 204 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd11
Genomic Map