Assay Details
Gene Symbol: Urgcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077661.1 | NP_001071129.1 | 4 - 5 | 646 | 81 | |
NM_178623.3 | NP_848738.2 | 4 - 5 | 230 | 81 | ||
GenBank mRNA | AK033577.1 | - | 4 - 5 | 204 | 81 | |
AK035580.1 | - | 4 - 5 | 642 | 81 | ||
AK041953.1 | - | 5 - 6 | 895 | 81 | ||
AK129378.1 | - | 3 - 4 | 288 | 81 | ||
BC066994.1 | - | 4 - 5 | 527 | 5696000 | 81 | |
BY058620.1 | - | 4 - 5 | 230 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
