Assay Details
Gene Symbol: Fbxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178624.6 | NP_848739.1 | 13 - 14 | 1006 | 73 | |
GenBank mRNA | AK039010.1 | - | 13 - 14 | 1006 | 73 | |
AK044693.1 | - | 13 - 14 | 1003 | 73 | ||
AK045742.1 | - | 13 - 14 | 993 | 73 | ||
AK089994.1 | - | 13 - 14 | 982 | 73 | ||
AK141177.1 | - | 13 - 14 | 996 | 73 | ||
AK163472.1 | - | 13 - 14 | 1002 | 73 | ||
BC096582.1 | - | 13 - 14 | 992 | 3963599 | 73 | |
BC145665.1 | - | 13 - 14 | 986 | 40130968 | 73 | |
BC145998.1 | - | 13 - 14 | 986 | 40131301 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
