Assay Details
Gene Symbol: Fbxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178624.6 | NP_848739.1 | 5 - 6 | 344 | 95 | |
GenBank mRNA | AK013142.1 | - | 6 - 7 | 478 | 95 | |
AK039010.1 | - | 5 - 6 | 344 | 95 | ||
AK044693.1 | - | 5 - 6 | 341 | 95 | ||
AK045742.1 | - | 5 - 6 | 331 | 95 | ||
AK089994.1 | - | 5 - 6 | 320 | 95 | ||
AK141177.1 | - | 5 - 6 | 334 | 95 | ||
AK163472.1 | - | 5 - 6 | 340 | 95 | ||
BC096582.1 | - | 5 - 6 | 330 | 3963599 | 95 | |
BC145665.1 | - | 5 - 6 | 324 | 40130968 | 95 | |
BC145998.1 | - | 5 - 6 | 324 | 40131301 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
