Assay Details
Gene Symbol: Fbxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178624.6 | NP_848739.1 | 7 - 8 | 512 | 80 | |
GenBank mRNA | AK013142.1 | - | 8 - 9 | 646 | 80 | |
AK039010.1 | - | 7 - 8 | 512 | 80 | ||
AK044693.1 | - | 7 - 8 | 509 | 80 | ||
AK045742.1 | - | 7 - 8 | 499 | 80 | ||
AK089994.1 | - | 7 - 8 | 488 | 80 | ||
AK141177.1 | - | 7 - 8 | 502 | 80 | ||
AK163472.1 | - | 7 - 8 | 508 | 80 | ||
BC051530.1 | - | 3 - 4 | 326 | 80 | ||
BC096582.1 | - | 7 - 8 | 498 | 3963599 | 80 | |
BC145665.1 | - | 7 - 8 | 492 | 40130968 | 80 | |
BC145998.1 | - | 7 - 8 | 492 | 40131301 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
