Assay Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178651.3 | NP_848766.2 | 12 - 13 | 1207 | 93 | |
| GenBank mRNA | AF263460.1 | - | 10 - 11 | 761 | 93 | |
| AK028885.1 | - | 12 - 13 | 1174 | 93 | ||
| AK033990.1 | - | 232 | 93 | |||
| AK035541.1 | - | 12 - 13 | 1194 | 93 | ||
| AK038559.1 | - | 11 - 12 | 1113 | 93 | ||
| AK082853.1 | - | 12 - 13 | 1209 | 93 | ||
| AK143277.1 | - | 12 - 13 | 1208 | 93 | ||
| AK179008.1 | - | 4 - 5 | 295 | 93 | ||
| AK182342.1 | - | 4 - 5 | 285 | 93 | ||
| AK204464.1 | - | 3 - 4 | 226 | 93 | ||
| AK205884.1 | - | 4 - 5 | 253 | 93 | ||
| AY682914.1 | - | 13 - 14 | 1200 | 93 | ||
| BC023833.1 | - | 4 - 5 | 311 | 93 | ||
| BC026565.1 | - | 8 - 9 | 625 | 93 | ||
| BC027806.1 | - | 12 - 13 | 969 | 93 | ||
| BC055773.1 | - | 9 - 10 | 660 | 93 | ||
| BC078440.1 | - | 10 - 11 | 784 | 93 | ||
| BC141008.1 | - | 13 - 14 | 1173 | 9055620 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a9
Genomic Map