Assay Details
| Gene Symbol: Slc30a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178651.3 | NP_848766.2 | 15 - 16 | 1557 | 83 | |
| GenBank mRNA | AF263460.1 | - | 13 - 14 | 1111 | 83 | |
| AK028885.1 | - | 15 - 16 | 1524 | 83 | ||
| AK033990.1 | - | 582 | 83 | |||
| AK035541.1 | - | 15 - 16 | 1544 | 83 | ||
| AK038559.1 | - | 14 - 15 | 1463 | 83 | ||
| AK082853.1 | - | 15 - 16 | 1559 | 83 | ||
| AK143277.1 | - | 15 - 16 | 1558 | 83 | ||
| AK149549.1 | - | 7 - 8 | 4008 | 83 | ||
| AY682914.1 | - | 16 - 17 | 1550 | 83 | ||
| BC023833.1 | - | 7 - 8 | 661 | 83 | ||
| BC024849.1 | - | 2 - 3 | 290 | 83 | ||
| BC026565.1 | - | 11 - 12 | 975 | 83 | ||
| BC027806.1 | - | 15 - 16 | 1319 | 83 | ||
| BC031705.1 | - | 12 - 13 | 1071 | 83 | ||
| BC055773.1 | - | 12 - 13 | 1010 | 83 | ||
| BC078440.1 | - | 13 - 14 | 1134 | 83 | ||
| BC141008.1 | - | 16 - 17 | 1523 | 9055620 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a9
Genomic Map