Assay Details
| Gene Symbol: B3gntl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178664.5 | NP_848779.2 | 10 - 11 | 1036 | 75 | |
| GenBank mRNA | AK031361.1 | - | 10 - 11 | 1025 | 75 | |
| AK156318.1 | - | 10 - 11 | 1014 | 75 | ||
| BC022739.1 | - | 4 - 5 | 348 | 75 | ||
| BC062106.1 | - | 4 - 5 | 895 | 75 | ||
| BC088993.1 | - | 10 - 11 | 901 | 6416623 | 75 | |
| BC125626.1 | - | 10 - 11 | 1009 | 40130141 | 75 | |
| BC132193.1 | - | 10 - 11 | 1009 | 40130470 | 75 | |
| BC145163.1 | - | 10 - 11 | 1013 | 75 | ||
| BC145165.1 | - | 10 - 11 | 1075 | 75 | ||
| BC145166.1 | - | 10 - 11 | 1009 | 9053717 | 75 | |
More Information
Set Membership: |
|

B3gntl1
Genomic Map