Assay Details
| Gene Symbol: B3gntl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178664.5 | NP_848779.2 | 4 - 5 | 448 | 76 | |
| GenBank mRNA | AK031361.1 | - | 4 - 5 | 437 | 76 | |
| AK043722.1 | - | 4 - 5 | 451 | 76 | ||
| AK156318.1 | - | 4 - 5 | 426 | 76 | ||
| BB641432.1 | - | 4 - 5 | 448 | 76 | ||
| BC125626.1 | - | 4 - 5 | 421 | 40130141 | 76 | |
| BC132193.1 | - | 4 - 5 | 421 | 40130470 | 76 | |
| BC145163.1 | - | 4 - 5 | 425 | 76 | ||
| BC145166.1 | - | 4 - 5 | 421 | 9053717 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

B3gntl1
Genomic Map