| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000003152.mm.1 |
NM_178675.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003152.mm.1 |
NM_178675.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003152.mm.1 |
TC1000000473.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000003150.mm.1 |
NM_178675.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003150.mm.1 |
NM_178675.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003150.mm.1 |
TC1000000473.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000003153.mm.1 |
NM_178675.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003153.mm.1 |
NM_178675.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003153.mm.1 |
TC1000000473.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000003151.mm.1 |
NM_178675.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003151.mm.1 |
NM_178675.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000003151.mm.1 |
TC1000000473.mm.1 |
MTA TC |