Assay Details
| Gene Symbol: Unc45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178680.4 | NP_848795.3 | 4 - 5 | 488 | 63 | |
| GenBank mRNA | AF539793.1 | - | 3 - 4 | 381 | 63 | |
| AK052294.1 | - | 4 - 5 | 415 | 63 | ||
| AK081821.1 | - | 4 - 5 | 427 | 63 | ||
| AK084418.1 | - | 4 - 5 | 456 | 63 | ||
| AK169274.1 | - | 4 - 5 | 427 | 63 | ||
| BC084585.1 | - | 4 - 5 | 421 | 30429260 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc45b
Genomic Map