Assay Details
| Gene Symbol: Dpy19l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178704.3 | NP_848819.2 | 4 - 5 | 564 | 119 | |
| GenBank mRNA | AF538322.1 | - | 4 - 5 | 548 | 119 | |
| AK031345.1 | - | 4 - 5 | 556 | 119 | ||
| AK033395.1 | - | 4 - 5 | 521 | 119 | ||
| AK034217.1 | - | 4 - 5 | 565 | 119 | ||
| AK045758.1 | - | 4 - 5 | 465 | 119 | ||
| BC050065.1 | - | 4 - 5 | 508 | 30016211 | 119 | |
| BC059897.1 | - | 4 - 5 | 461 | 6821433 | 119 | |
More Information
Set Membership: |
|

Dpy19l3
Genomic Map