Assay Details
Gene Symbol: Siglech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178706.4 | NP_848821.2 | 1 - 2 | 545 | 89 | |
GenBank mRNA | AK032377.1 | - | 1 - 2 | 498 | 89 | |
AK045719.1 | - | 2 - 3 | 175 | 89 | ||
AK139362.1 | - | 2 - 3 | 446 | 89 | ||
BC098229.1 | - | 1 - 2 | 484 | 1430130 | 89 | |
BC120501.1 | - | 1 - 2 | 487 | 40129424 | 89 | |
BC144742.1 | - | 1 - 2 | 487 | 9053290 | 89 | |
BC144743.1 | - | 1 - 2 | 487 | 9053291 | 89 | |
DQ157764.1 | - | 1 - 2 | 464 | 89 | ||
DQ208935.1 | - | 1 - 2 | 490 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK045719.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
