Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800007346.mm.1 |
NM_001289666.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007346.mm.1 |
NM_001289667.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007346.mm.1 |
NM_178801.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007346.mm.1 |
NR_110356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007346.mm.1 |
TC1800000990.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800007344.mm.1 |
NM_001289666.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007344.mm.1 |
NM_001289667.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007344.mm.1 |
NM_178801.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007344.mm.1 |
NR_110356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800007344.mm.1 |
TC1800000990.mm.1 |
MTA TC |