Assay Details
| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178874.2 | NP_849205.1 | 1 - 2 | 302 | 66 | |
| GenBank mRNA | AK172957.1 | - | 1 - 2 | 157 | 66 | |
| BC052035.1 | - | 1 - 2 | 302 | 5695005 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmcc2
Genomic Map