Assay Details
Gene Symbol: Mtmr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181409.3 | NP_852074.2 | 6 - 7 | 784 | 79 | |
GenBank mRNA | AK132375.1 | - | 6 - 7 | 751 | 79 | |
BC051083.1 | - | 6 - 7 | 717 | 6486043 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
