Assay Details
| Gene Symbol: Gtf2h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181410.3 | NP_852075.1 | 6 - 7 | 493 | 86 | |
| GenBank mRNA | AK050227.1 | - | 6 - 7 | 477 | 86 | |
| AK133941.1 | - | 6 - 7 | 464 | 86 | ||
| BC017515.1 | - | 6 - 7 | 470 | 4504077 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2h3
Genomic Map