Assay Details
Gene Symbol: Gtf2h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181410.3 | NP_852075.1 | 7 - 8 | 532 | 123 | |
GenBank mRNA | AK050227.1 | - | 7 - 8 | 516 | 123 | |
AK133941.1 | - | 7 - 8 | 503 | 123 | ||
BC017515.1 | - | 7 - 8 | 509 | 4504077 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
