Assay Details
Gene Symbol: Gtf2h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181410.3 | NP_852075.1 | 9 - 10 | 656 | 75 | |
GenBank mRNA | AK133941.1 | - | 9 - 10 | 627 | 75 | |
BC017515.1 | - | 9 - 10 | 633 | 4504077 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
