Assay Details
| Gene Symbol: Pik3c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181414.5 | NP_852079.2 | 4 - 5 | 586 | 69 | |
| GenBank mRNA | AK030578.1 | - | 4 - 5 | 616 | 69 | |
| AK044997.1 | - | 4 - 5 | 586 | 69 | ||
| BC024675.1 | - | 4 - 5 | 621 | 3661094 | 69 | |
| BC027029.1 | - | 4 - 5 | 555 | 69 | ||
| BC057678.1 | - | 4 - 5 | 589 | 3496633 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3c3
Genomic Map