Assay Details
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083328.1 | NP_001076797.1 | 8 - 9 | 1037 | 104 | |
NM_001083329.1 | NP_001076798.1 | 7 - 8 | 943 | 104 | ||
NM_028422.1 | NP_082698.1 | 8 - 9 | 1082 | 104 | ||
NM_181424.3 | NP_852089.2 | 9 - 10 | 1176 | 104 | ||
GenBank mRNA | AK007444.1 | - | 6 - 7 | 1028 | 104 | |
AK040689.1 | - | 8 - 9 | 1566 | 104 | ||
BC025144.1 | - | 9 - 10 | 1205 | 4483063 | 104 | |
BC028465.1 | - | 3 - 4 | 478 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901244 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901243 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901245 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901246 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050807 | BC039784.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050807 | BC025144.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050807 | BC028465.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050807 | BC028518.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050807 | BC030918.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640052 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351763 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687563 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418426 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795270 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970538 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93772 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95240 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459345 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177554 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 78328 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106773 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161506 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133604 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119211 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050807 | BC039784.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050807 | BC025144.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050807 | BC028465.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050807 | BC028518.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050807 | BC030918.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486906 | BC039784.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486906 | BC025144.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486906 | BC028465.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486906 | BC028518.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P486906 | BC030918.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901246 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901248 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901255 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901258 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901251 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901260 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21901261 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119211 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106773 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161506 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133604 | - | - |
Whole Mouse Genome G4122A | A_51_P486906 | BC039784.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P486906 | BC025144.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P486906 | BC028465.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P486906 | BC028518.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P486906 | BC030918.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131823 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134641 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119211 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133604 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106773 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161506 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901246 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901243 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901244 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21901245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21901359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01680704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901359 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01979038 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02172095 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02041038 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01634522 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901275 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02049784 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02041029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901307 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01501726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01261125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01217094 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01979041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02238293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901308 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01979045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02041036 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901368 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901331 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901349 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01412823 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901270 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01483314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02049778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901343 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901348 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901379 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01979046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901365 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01680697 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901252 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02053564 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02223735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901328 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01680713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02041037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901362 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901366 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901324 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01261123 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21901332 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01696546 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02049781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01412821 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01979043 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10479326 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10479325 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10479328 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10479334 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10479331 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10479333 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10479329 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10479330 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10479327 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10479332 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451467_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452636_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1430272_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10479324 | - | - |
Mouse Genome 430 2.0 Array | 1452636_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1460029_at | - | - |
Mouse Genome 430 2.0 Array | 1451467_s_at | - | - |
Mouse Genome 430 2.0 Array | 1460029_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1451467_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1452636_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52856_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52856_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200022712.mm.1 | NM_001083328.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022712.mm.1 | NM_001083328.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022712.mm.1 | NM_001083329.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022712.mm.1 | NM_001083329.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022712.mm.1 | NM_028422.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022712.mm.1 | NM_028422.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022712.mm.1 | NM_181424.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022712.mm.1 | NM_181424.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022712.mm.1 | TC0200002781.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200022715.mm.1 | NM_001083328.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022715.mm.1 | NM_001083328.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022715.mm.1 | NM_001083329.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022715.mm.1 | NM_001083329.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022715.mm.1 | NM_028422.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022715.mm.1 | NM_028422.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022715.mm.1 | NM_181424.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022715.mm.1 | NM_181424.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200022715.mm.1 | TC0200002781.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200022709.mm.1 | TC0200002781.mm.1 | MTA TC |
Mouse Expression Set 430 | 1451467_PM_s_at | - | - |
Mouse Expression Set 430 | 1452636_PM_x_at | - | - |
Mouse Expression Set 430 | 1451467_s_at | - | - |
Mouse Expression Set 430 | 1452636_x_at | - | - |
Mouse Expression Set 430 | 1460029_at | - | - |
Mouse Expression Set 430 | 1460029_PM_at | - | - |
Murine Genome U74v2 Set | 130778_PM_at | - | - |
Murine Genome U74v2 Set | 130778_at | - | - |
Mouse Genome 430A 2.0 Array | 1452636_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1452636_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1451467_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1451467_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452636_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451467_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452636_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451467_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430272_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430272_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3060609 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003060609 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2759167 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2759167 | - | - |
mouse-6 v1.1 (Illumina) | scl052856.8_265-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3060609 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl052856.8_265-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060609 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2759167 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2759167 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl052856.8_265-S | - | - |
