Assay Details
| Gene Symbol: Ehhadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023737.3 | NP_076226.2 | 5 - 6 | 618 | 89 | |
| GenBank mRNA | AK004867.1 | - | 5 - 6 | 589 | 89 | |
| BC016899.1 | - | 5 - 6 | 592 | 4241677 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ehhadh
Genomic Map