Assay Details
| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025391.2 | NP_079667.2 | 3 - 4 | 336 | 67 | |
| NR_028367.1 | 2 - 3 | 429 | 67 | |||
| GenBank mRNA | AK003739.1 | - | 3 - 4 | 336 | 67 | |
| AK006057.1 | - | 3 - 4 | 316 | 67 | ||
| AK014290.1 | - | 3 - 4 | 295 | 67 | ||
| BC003972.1 | - | 3 - 4 | 323 | 3488726 | 67 | |
| BE623972.1 | - | 2 - 3 | 400 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nip7
Genomic Map