Assay Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025505.4 | NP_079781.2 | 2 - 3 | 250 | 120 | |
| GenBank mRNA | AK029787.1 | - | 2 - 3 | 251 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029787.1, NM_025505.4 transcripts.
Set Membership: |
|

Blzf1
Genomic Map