Assay Details
Gene Symbol: Suox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173733.3 | NP_776094.2 | 3 - 4 | 305 | 72 | |
GenBank mRNA | AK149422.1 | - | 3 - 4 | 293 | 72 | |
AK154608.1 | - | 3 - 4 | 293 | 72 | ||
BC027197.1 | - | 2 - 3 | 260 | 4160277 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542560 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542598 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542559 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542562 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542596 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542561 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542597 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542563 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249360 | BC027197.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249360 | AK154608.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249360 | NM_173733.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249360 | AK149422.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249360 | BC027197.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249360 | AK154608.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249360 | NM_173733.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249360 | AK149422.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684471 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833885 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115123 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249360 | BC027197.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249360 | AK154608.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249360 | NM_173733.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249360 | AK149422.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249360 | BC027197.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249360 | AK154608.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249360 | NM_173733.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249360 | AK149422.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249360 | BC027197.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249360 | AK154608.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249360 | NM_173733.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249360 | AK149422.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249360 | BC027197.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249360 | AK154608.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249360 | NM_173733.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249360 | AK149422.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542577 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542580 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542589 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542596 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542600 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542597 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542605 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542583 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542578 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542581 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542620 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542603 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542576 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542573 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542575 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542595 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542584 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542593 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542571 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542587 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542586 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542569 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542598 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27542568 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115123 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07233797 | - | - |
Whole Mouse Genome G4122A | A_51_P249360 | BC027197.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P249360 | AK154608.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P249360 | NM_173733.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P249360 | AK149422.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P249360 | BC027197.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P249360 | AK154608.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P249360 | NM_173733.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P249360 | AK149422.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108670 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115123 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08821051 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08409700 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08821050 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08409699 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08821027 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542598 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542559 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542563 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542562 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542597 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542561 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542560 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27542596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27542540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08968116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08840794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08409698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08821039 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542580 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542603 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08377570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542531 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542601 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542598 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08819218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542583 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08458506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08377573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08868927 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08377576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542577 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08821033 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08868929 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08377571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08748788 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08215906 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08453172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542575 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08295194 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27542618 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10373517 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373518 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10373516 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451339_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10373515 | - | - |
Mouse Genome 430 2.0 Array | 1451339_at | - | - |
Mouse Genome 430 2.0 Array | 1451339_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 211389_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 211389_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000023791.mm.1 | NM_173733.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000023791.mm.1 | NM_173733.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000023791.mm.1 | TC1000003158.mm.1 | MTA TC |
Mouse Expression Set 430 | 1451339_at | - | - |
Mouse Expression Set 430 | 1451339_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451339_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451339_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451339_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451339_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 637629 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5220286 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005220286 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246916 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1246916 | - | - |
mouse-6 v1.1 (Illumina) | scl37349.4_206-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5220286 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl37349.4_206-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5220286 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2912532 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2912532 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37349.4_206-S | - | - |
