Assay Details
| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110496.1 | NP_001103966.1 | 5 - 6 | 532 | 93 | |
| NM_001110497.1 | NP_001103967.1 | 5 - 6 | 532 | 93 | ||
| NM_173734.3 | NP_776095.2 | 5 - 6 | 532 | 93 | ||
| GenBank mRNA | AK044591.1 | - | 5 - 6 | 486 | 93 | |
| AK086738.1 | - | 5 - 6 | 482 | 93 | ||
| AK134268.1 | - | 5 - 6 | 495 | 93 | ||
| AK137137.1 | - | 5 - 6 | 498 | 93 | ||
| BC027354.1 | - | 5 - 6 | 505 | 4010477 | 93 | |
More Information
Set Membership: |
|

Tmem87a
Genomic Map