Assay Details
| Gene Symbol: Wdr24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173741.3 | NP_776102.1 | 2 - 3 | 1242 | 84 | |
| GenBank mRNA | AK078694.1 | - | 2 - 3 | 1222 | 84 | |
| AK170253.1 | - | 2 - 3 | 1222 | 84 | ||
| BC037651.1 | - | 2 - 3 | 1164 | 5148210 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr24
Genomic Map