Assay Details
| Gene Symbol: Ilvbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173751.4 | NP_776112.1 | 12 - 13 | 1651 | 92 | |
| GenBank mRNA | AK087956.1 | - | 11 - 12 | 1525 | 92 | |
| AK146075.1 | - | 11 - 12 | 1579 | 92 | ||
| BC052054.1 | - | 11 - 12 | 1985 | 92 | ||
| BC056459.1 | - | 12 - 13 | 1609 | 5718157 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ilvbl
Genomic Map