Assay Details
Gene Symbol: Ilvbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173751.4 | NP_776112.1 | 7 - 8 | 993 | 63 | |
GenBank mRNA | AK030933.1 | - | 4 - 5 | 431 | 63 | |
AK087956.1 | - | 6 - 7 | 867 | 63 | ||
AK133553.1 | - | 6 - 7 | 923 | 63 | ||
AK138031.1 | - | 6 - 7 | 844 | 63 | ||
AK143918.1 | - | 6 - 7 | 892 | 63 | ||
AK146075.1 | - | 6 - 7 | 921 | 63 | ||
AK169899.1 | - | 6 - 7 | 919 | 63 | ||
BC052054.1 | - | 6 - 7 | 1327 | 63 | ||
BC056459.1 | - | 7 - 8 | 951 | 5718157 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
