Assay Details
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291049.1 | NP_001277978.1 | 13 - 14 | 2258 | 91 | |
NM_173754.4 | NP_776115.2 | 13 - 14 | 2273 | 91 | ||
GenBank mRNA | AK042217.1 | - | 13 - 14 | 2259 | 91 | |
AK047339.1 | - | 1 - 2 | 246 | 91 | ||
AK083271.1 | - | 13 - 14 | 2111 | 91 | ||
BC021474.1 | - | 12 - 13 | 1563 | 5343429 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK047339.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
