Assay Details
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291049.1 | NP_001277978.1 | 5 - 6 | 1072 | 82 | |
NM_173754.4 | NP_776115.2 | 5 - 6 | 1072 | 82 | ||
GenBank mRNA | AK042217.1 | - | 5 - 6 | 1073 | 82 | |
AK083271.1 | - | 5 - 6 | 910 | 82 | ||
BC021474.1 | - | 4 - 5 | 362 | 5343429 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC021474.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
