Assay Details
Gene Symbol: Ccdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK048790.1 | - | 4 - 5 | 1739 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK048790.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
