Assay Details
| Gene Symbol: Galm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176963.4 | NP_795937.1 | 2 - 3 | 587 | 70 | |
| GenBank mRNA | AK040998.1 | - | 2 - 3 | 588 | 70 | |
| AK158120.1 | - | 2 - 3 | 386 | 70 | ||
| BC028818.1 | - | 2 - 3 | 390 | 5135570 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galm
Genomic Map