Assay Details
| Gene Symbol: Nt5dc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176968.4 | NP_795942.2 | 1 - 2 | 173 | 94 | |
| GenBank mRNA | AK049656.1 | - | 1 - 2 | 173 | 94 | |
| AK077876.1 | - | 1 - 2 | 174 | 94 | ||
| AK145666.1 | - | 1 - 2 | 174 | 94 | ||
| AK161350.1 | - | 1 - 2 | 127 | 94 | ||
| AK164144.1 | - | 1 - 2 | 117 | 94 | ||
| AK171761.1 | - | 1 - 2 | 186 | 94 | ||
| BC125276.1 | - | 1 - 2 | 172 | 40107511 | 94 | |
More Information
Set Membership: |
|

Nt5dc1
Genomic Map