Assay Details
| Gene Symbol: Nt5dc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176968.4 | NP_795942.2 | 3 - 4 | 339 | 62 | |
| GenBank mRNA | AK049656.1 | - | 3 - 4 | 339 | 62 | |
| AK077876.1 | - | 3 - 4 | 340 | 62 | ||
| AK145666.1 | - | 3 - 4 | 340 | 62 | ||
| AK161350.1 | - | 3 - 4 | 293 | 62 | ||
| AK164144.1 | - | 3 - 4 | 283 | 62 | ||
| AK171761.1 | - | 3 - 4 | 352 | 62 | ||
| BC098476.1 | - | 3 - 4 | 302 | 30051251 | 62 | |
| BC125276.1 | - | 3 - 4 | 338 | 40107511 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nt5dc1
Genomic Map