| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400011565.mm.1 |
NM_030180.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011565.mm.1 |
NM_030180.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011565.mm.1 |
TC1400001544.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400011566.mm.1 |
NM_030180.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011566.mm.1 |
NM_030180.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011566.mm.1 |
TC1400001544.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400011569.mm.1 |
NM_030180.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011569.mm.1 |
NM_030180.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011569.mm.1 |
TC1400001544.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400011568.mm.1 |
NM_030180.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011568.mm.1 |
NM_030180.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011568.mm.1 |
TC1400001544.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400011570.mm.1 |
NM_030180.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011570.mm.1 |
NM_030180.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400011570.mm.1 |
TC1400001544.mm.1 |
MTA TC |