Assay Details
| Gene Symbol: Gnal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010307.3 | NP_034437.1 | 3 - 4 | 534 | 113 | |
| NM_177137.5 | NP_796111.2 | 3 - 4 | 699 | 113 | ||
| GenBank mRNA | AK032407.1 | - | 3 - 4 | 345 | 113 | |
| AK035952.1 | - | 3 - 4 | 700 | 113 | ||
| AK045484.1 | - | 3 - 4 | 664 | 113 | ||
| AK158831.1 | - | 3 - 4 | 535 | 113 | ||
| AY179169.1 | - | 3 - 4 | 268 | 113 | ||
| BC078439.1 | - | 3 - 4 | 661 | 5685967 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnal
Genomic Map