Assay Details
| Gene Symbol: Svopl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177200.4 | NP_796174.2 | 8 - 9 | 767 | 64 | |
| GenBank mRNA | AK034997.1 | - | 8 - 9 | 768 | 64 | |
| AK137043.1 | - | 8 - 9 | 768 | 64 | ||
| BC058741.1 | - | 8 - 9 | 750 | 6309979 | 64 | |
More Information
Set Membership: |
|

Svopl
Genomic Map