Assay Details
| Gene Symbol: Bend6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177235.3 | NP_796209.2 | 5 - 6 | 854 | 97 | |
| GenBank mRNA | AK044633.1 | - | 4 - 5 | 793 | 97 | |
| AK134655.1 | - | 5 - 6 | 997 | 97 | ||
| BC057378.1 | - | 5 - 6 | 854 | 5707039 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bend6
Genomic Map