Assay Details
| Gene Symbol: Dcaf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177267.4 | NP_796241.3 | 2 - 3 | 556 | 77 | |
| GenBank mRNA | AK122561.1 | - | 2 - 3 | 393 | 77 | |
| AK137579.1 | - | 2 - 3 | 556 | 77 | ||
| AK165096.1 | - | 2 - 3 | 540 | 77 | ||
| BC042567.1 | - | 2 - 3 | 230 | 77 | ||
| BC046813.1 | - | 2 - 3 | 494 | 5702325 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dcaf5
Genomic Map