Assay Details
Gene Symbol: Serac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111017.1 | NP_001104487.1 | 10 - 11 | 1140 | 83 | |
NM_177311.4 | NP_796285.2 | 9 - 10 | 1050 | 83 | ||
GenBank mRNA | AK019677.1 | - | 9 - 10 | 1039 | 83 | |
AK154605.1 | - | 10 - 11 | 1091 | 83 | ||
AK155571.1 | - | 10 - 11 | 1143 | 83 | ||
AK170498.1 | - | 10 - 11 | 1091 | 83 | ||
BC119594.1 | - | 10 - 11 | 1129 | 40059066 | 83 | |
BC119595.1 | - | 10 - 11 | 1129 | 40059071 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
