Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600020267.mm.1 |
TC0600002610.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020267.mm.1 |
NM_001003955.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020267.mm.1 |
NM_001003955.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020267.mm.1 |
NM_177466.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020267.mm.1 |
NM_177466.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020266.mm.1 |
TC0600002610.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020266.mm.1 |
NM_001003955.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020266.mm.1 |
NM_001003955.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020266.mm.1 |
NM_177466.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020266.mm.1 |
NM_177466.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020268.mm.1 |
TC0600002610.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600020268.mm.1 |
NM_001003955.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020268.mm.1 |
NM_001003955.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020268.mm.1 |
NM_177466.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600020268.mm.1 |
NM_177466.4 |
Refseq |