Assay Details
| Gene Symbol: Acaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177470.3 | NP_803421.1 | 5 - 6 | 618 | 75 | |
| GenBank mRNA | AK002555.1 | - | 5 - 6 | 608 | 75 | |
| AK050101.1 | - | 5 - 6 | 620 | 75 | ||
| AK146007.1 | - | 5 - 6 | 633 | 75 | ||
| AK167567.1 | - | 5 - 6 | 619 | 75 | ||
| AK167715.1 | - | 5 - 6 | 616 | 75 | ||
| AK169359.1 | - | 5 - 6 | 619 | 75 | ||
| BC028901.1 | - | 5 - 6 | 613 | 4195488 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acaa2
Genomic Map