Assay Details
| Gene Symbol: Usp46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177561.3 | NP_808229.1 | 5 - 6 | 766 | 125 | |
| GenBank mRNA | AK029604.1 | - | 4 - 5 | 618 | 125 | |
| AK145298.1 | - | 5 - 6 | 786 | 125 | ||
| BC030888.1 | - | 3 - 4 | 533 | 125 | ||
| BC039916.1 | - | 5 - 6 | 768 | 4950034 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp46
Genomic Map