Assay Details
Gene Symbol: 6820408C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289738.1 | NP_001276667.1 | 5 - 6 | 709 | 60 | |
NM_177656.4 | NP_808324.1 | 5 - 6 | 706 | 60 | ||
NR_110367.1 | 5 - 6 | 706 | 60 | |||
NR_110368.1 | 5 - 6 | 706 | 60 | |||
GenBank mRNA | AK078482.1 | - | 5 - 6 | 706 | 60 | |
BC137826.1 | - | 4 - 5 | 586 | 8860845 | 60 | |
BC144943.1 | - | 4 - 5 | 583 | 60 | ||
BC144944.1 | - | 4 - 5 | 586 | 9053492 | 60 | |
BC144945.1 | - | 4 - 5 | 579 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344942 | BC144944.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344942 | BC137826.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344942 | NM_177656.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344942 | AK078482.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344942 | BC144944.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344942 | BC137826.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344942 | NM_177656.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P344942 | AK078482.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194023 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61335 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156244 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359520 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727289 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898713 | BC116754.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898713 | BC116756.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898713 | NM_147108.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43545 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168683 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822527 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398019 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758003 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972175 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304203 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92861 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10924 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168226 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159729 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344942 | BC144944.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344942 | BC137826.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344942 | NM_177656.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344942 | AK078482.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344942 | BC144944.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344942 | BC137826.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344942 | NM_177656.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P344942 | AK078482.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749247 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749235 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749233 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749236 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749238 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21749239 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00528508 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168226 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04741841 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159729 | - | - |
Whole Mouse Genome G4122A | A_51_P344942 | BC144944.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344942 | BC137826.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344942 | NM_177656.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344942 | AK078482.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344942 | BC144944.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344942 | BC137826.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344942 | NM_177656.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P344942 | AK078482.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106603 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111742 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159729 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168226 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01318128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21749297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01655458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749328 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01318127 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749320 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02196289 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749290 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749323 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01930282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02282069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01371061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01930272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21749330 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02267720 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10477116 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477111 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477109 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477119 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477117 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477110 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477108 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477115 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477118 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477114 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477112 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477113 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1441306_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10477107 | - | - |
Mouse Genome 430 2.0 Array | 1441306_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1442366_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1441306_at | - | - |
Mouse Genome 430 2.0 Array | 1442366_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228778_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228778_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200017880.mm.1 | NM_001289738.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017880.mm.1 | NM_177656.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017880.mm.1 | NR_110367.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017880.mm.1 | NR_110368.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017880.mm.1 | TC0200002282.mm.1 | MTA TC |
Mouse Expression Set 430 | 1441306_PM_at | - | - |
Mouse Expression Set 430 | 1441306_at | - | - |
Murine Genome U74v2 Set | 129855_PM_g_at | - | - |
Murine Genome U74v2 Set | 129854_PM_at | - | - |
Murine Genome U74v2 Set | 129855_g_at | - | - |
Murine Genome U74v2 Set | 129854_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441306_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441306_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 651224 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4730112 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004730112 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245721 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1245721 | - | - |
mouse-6 v1.1 (Illumina) | scl20121.12.1_122-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4730112 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl20121.12.1_122-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4730112 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1245721 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245721 | - | - |
